Learn More
Haldenwanger™ Aluminum Oxide Combustion Boats
Product Code. 12317018
Change view
Click to view available options
Dimensions (L x W x H):
115 x 16 x 9 mm
120 x 30 x 15 mm
160 x 40 x 21 mm
200 x 15 x 15 mm
45 x 11 x 7 mm
75 X 11 X 7 mm
87 x 17 x 9 mm
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
12317018 75 X 11 X 7 mm Each
12337018 115 x 16 x 9 mm Each
12347018 120 x 30 x 15 mm Each
12357018 160 x 40 x 21 mm Each
12367018 200 x 15 x 15 mm Each
12397008 45 x 11 x 7 mm Each
12327018 87 x 17 x 9 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12317018 Supplier Haldenwanger™ Supplier No. L ALS.SCH.75X11X7
  • ONLINE EXCLUSIVE PRICE
    50.95€
    45.28€ / Pack of 1
    Save 5.67€
    11% Off
Estimated Shipment: 28-09-2026
Use these boats for a variety laboratory applications. Haldenwanger™ Aluminum Oxide Combustion Boats are ideal for chemical calcining, metal casting and sample preparation.
Aluminum oxide boats of great hardness, strength and chemical resistance. Suitable for continuous use up to 1, 700°C to 1, 750°C.
Specifications
Type Combustion Boat
Dimensions (L x W x H) 75 X 11 X 7 mm
For Use With (Application) For continuous use up to 1700 to 1750°C.
Height (Metric) 7 mm
Material Aluminum Oxide
Quantity 1 Ea.