Learn More
Brady™ Aluminum Mandatory Signs
Specifications
Specifications
| Certifications/Compliance | NF X 08-003-1 and 08-003-3 NX and the European Directive 92/58 / EEC |
| Material | Aluminum |
| Quantity | 1 Ea. |
| Diameter (Metric) | 100 mm |
| For Use With (Application) | Indoor and outdoor safety signs |
| Shape | Round |
| Displayed Text | Cotton Blouse |
| Product Type | Mandatory Sign |
| Color | White on Blue |