Learn More
Welch™ Aluminum KF Inner Centering with Viton O-ring
Product Code. 10680343 Shop All Welch Ilmvac Products
Change view
Click to view available options
For Use With:
Separator 16 DN
Separator 25 DN
Separator 40 DN
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Unit Size
10680343 Separator 16 DN Each
10152522 Separator 25 DN Each
10090992 Separator 40 DN Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10680343 Supplier Welch™ Supplier No. 701091
  • ONLINE EXCLUSIVE PRICE
    7.15€
    6.77€ / Pack of 1
    Save 0.38€
    5% Off
Estimated Shipment: 04-11-2026
Designed for use with CRVpro Rotary Vane Vacuum Pumps. Welch Ilmvac™ Aluminum KF Inner Centering Rings feature a replaceable Fluoroelastomer O-ring seal.
  • Stainless steel centering ring with Viton O-ring
  • Replaceable Fluoroelastomer O-ring seal
  • Centering ring assemblies are placed between two ISO flanges with matching outer diameters
  • he widest portion of the centering ring rests inside a centering ring locater groove (blank flange) or counterbore (bored flange) on the Kwik-Flange™
  • The O-ring rests on the flat polished surface outside the locater groove or counterbore
TRUSTED_SUSTAINABILITY
Specifications
For Use With Separator 16 DN
Material Aluminum
Quantity 1 Each
Type KF Inner Centering