Learn More
Agencinox™ Aluminum Instrument Box
Product Code. 11812682
Change view
Click to view available options
Color:
Blue
Yellow
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11812682 Yellow Each
11892672 Blue Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11812682 Supplier Agencinox™ Supplier No. 8030J
  • ONLINE EXCLUSIVE PRICE
    38.02€
    18.74€ / Pack of 1
    Save 19.28€
    50% Off
Estimated Shipment: 02-12-2026
Colored, anodized aluminum box suitable for instrument sterilization and storage.
  • Available in different colors
  • Anodized aluminum construction
  • Suitable for sterilization and storage of instruments
Specifications
Color Yellow
Height (Metric) Exterior 30 mm
Length (Metric) 180 mm
Material Anodized Aluminum
Width (Metric) Exterior 90 mm
Dimensions (L x W x H) 180 x 90 x 30 mm