Learn More
New Brunswick Scientific™ Erlenmeyer Flask Dedicated Platform
Click to view available options
For Use With (Equipment):
Innova 44/44R Shakers
Unit Size:
Each
Description
- 2L Erlenmeyer flask dedicated platform
- Heavy duty 0.95cm thick aluminum
- For Innova 44/44R Incubator Shakers
- 76 x 46cm
- Not interchangeable with other 76 x 46cm platforms
Specifications
Specifications
| Product Type | Erlenmeyer Flask Dedicated Platform |
| Material | Aluminum |
| Dimensions (L x W) | 76 x 46 cm |
| Thickness (Metric) | 0.95 cm |
| For Use With (Equipment) | Innova 44/44R Shakers |