Learn More
Buerkle™ Aluminum EasyScoop™
Product Code. p-8001764 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
1500 mL
2000 mL
Length (Metric):
200 mm
300 mm
Unit Size:
Each
Product Code. 10458023
Brand: Buerkle™ 53561500
Aluminum EasyScoop™ with a centrally arranged inner grip that is ergonomically designed to ensure easy and safe work.
  • Considerably shorter than conventional scoops, it is easier to take samples from narrow containers.
  • Made of light aluminum, bright-anodised
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1500 mL
Product Type Measuring Scoop
Finish Bright-anodised
For Use With (Application) Used to Take Samples From Narrow or Confined Containers
Material Aluminum
Handle Type Ergonomic
Length (Metric) 200 mm