Learn More
KGW Isotherm™ Dish-Shaped Dewar Flasks
Product Code. 10139700
Change view
Click to view available options
Capacity (Metric):
260 mL
400 mL
800 mL
5300 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10139700 400 mL Each
10790972 800 mL Each
10458551 5300 mL Each
10221461 260 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10139700 Supplier KGW Isotherm™ Supplier No. 1194
  • ONLINE EXCLUSIVE PRICE
    422.05€
    269.31€ / Pack of 1
    Save 152.74€
    36% Off
View all promotions
Estimated Shipment: 25-11-2026
For the cooling of small samples. KGW Isotherm™ Dish-Shaped Dewar Flasks are suitable as a hot or cold bath.
  • Easy handling
  • Suitable on magnetic stirrer
  • Full silvered
  • Made of borosilicate glass 3.3
  • Polyethylene lid (not included)
Specifications
Color Silver
Diameter (Metric) Outer 130mm
For Use With (Application) Laboratory technique, Medicinal technique
Height (Metric) Inner 70mm
Height (Metric) 115mm
Base Material Borosilicate Glass
Capacity (Metric) 400 mL
Diameter (Metric) Inner 110 mm
Material Aluminum