Learn More
Sperian™ Honeywell™ Aluminum RD40 Filters
Click to view available options
Model:
Model A2B2E2K2RD
Model A2B2P3RD
Model ABEK2P3RD
Model AXB2P3
Model HgP3
Tamaño de la unidad:
Each
Descripción
- Body made of aluminum provides optimum resistance to hazards
- Low breathing resistance
- Available in models to protect against airborne particulates, biohazards, chemicals, contamination and gas, vapors and smoke
Especificaciones
Especificaciones
| Cartridge Type | Canister |
| Certifications/Compliance | E.C. Declaration of Conformity- EC Category PPE- 3, Quality Assurance- ISO 9001 / 2000, EC Certificate Number- 7752 A/13/1 PSA, EC Attestation- EC Attestation, EC Attestation Number- 7752 A/13/1 PSA, Certifications- EN 14387, EN 143, EN 12941, HON filter |
| Connection Type | RD40 |
| Material | Aluminum alloy with PE cover activated and impregnated charcoal. |
| Product Type | ALU RD40 A2B2P3 |
| For Use With (Application) | Respiratory protection against organic gases and vapors with a boiling point below 65°C, inorganic gases and vapors, solid and liquid particles |
| For Use With (Equipment) | Full-Face mask equipped with an Rd40 connector. |
| Model | Model A2B2P3RD |
| Quantity | 1 Ea. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido