Learn More
Horst™ Aluminum Blanks
Product Code. 10382362
Change view
Click to view available options
Capacity (Metric):
500 mL
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10382362 500 mL Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10382362 Supplier Horst™ Supplier No. 080615
  • ONLINE EXCLUSIVE PRICE
    36.96€
    23.11€ / Pack of 25
    Save 13.85€
    37% Off
Estimated Shipment: 13-01-2027
Specifications
Capacity (Metric) 500 mL
Product Type Aluminum Blanks
For Use With (Application) For heating mantles