Learn More
Brady™ Blank Safety Lockout Hasps
Product Code. 15633527
Click to view available options
Color:
Green
Red
Silver
Unit Size:
1 pack
Pack of 5
Product Code. 15633527
Brand: Brady™ 265399
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Combine these blank hasps with your own customized labels.
  • Aluminum
  • 6 padlocks can be applied
Specifications
Product Type Blank Safety Lockout Hasps
Alternate Part Number 265399
Color Red
Material Aluminum
Depth (Metric) 4.7 mm
Diameter (Metric) Shackle 8 mm
For Use With (Application) For Blank Safety Lockout Hasps
Height (Metric) 184.1 mm
Quantity 5 Pk.
Temperature Range (Metric) −30° to +175°C
Width (Metric) 76.2 mm