Learn More
Nickel Electro™ Aluminum Basin with Lid
Product Code. p-8013866 Shop All Nickel-Electro Products
Click to view available options
Capacity (Metric):
8 mL
13 mL
45 mL
Unit Size:
Each
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Product Code. 12747356
Brand: Nickel Electro™ 650
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Made of durable aluminum, these Nickel Electro™ Aluminum Basins with Lid are ideal in laboratory environments.
  • For coal moisture determination
  • Flat, bottom aluminum basin with lid 
Specifications
Bottom Shape Flat
Capacity (Metric) 45 mL
Material Aluminum
Type Evaporating Basin
For Use With (Application) For Coal Moisture Determination
Height (Metric) 14 mm
Diameter (Metric) 65 mm