missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum, Al/Si/Cu, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Aluminum, Al/Si/Cu, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Nickel/Vanadium/Manganese/Cobalt/Bismuth/Chromium/Titanium/Lead/Magnesium/Zinc/Iron/Copper/Silicon/Tin |
| CAS | 7429-90-5 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Nickel 0.012 %/Vanadium 0.012 %/Manganese 0.015 %/Cobalt 0.022 %/Bismuth 0.07 %/Chromium 0.2 %/Titanium 0.21 %/Lead 0.24 %/Magnesium 0.29 %/Zinc 1.14 %/Iron 1.78 %/Copper 4.34 %/Silicon 7.69 %/Tin (0.008) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction