Learn More
Masterflex™ Aluminum 16-Turn Valve Cartridge
Product Code. 15207228
Change view
Click to view available options
Airflow Rate:
0 to 200 mL/min.
1001 to 2500 mL/min.
6201 mL/min.
Waterflow Rate:
0 to 6 mL/min.
30 to 70 mL/min.
650 mL/min.
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Airflow Rate Waterflow Rate unitSize
15207228 1001 to 2500 mL/min. 30 to 70 mL/min. Each
15297218 0 to 200 mL/min. 0 to 6 mL/min. Each
15334384 6201 mL/min. 650 mL/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15207228 Supplier Masterflex™ Supplier No. 0321862
  • ONLINE EXCLUSIVE PRICE
    140.00€
    108.53€ / Pack of 1
    Save 31.47€
    22% Off
Estimated Shipment: 11-09-2026
Specifications
Description Range 4
For Use With (Equipment) Aluminum Frame Systems
Type 16-Turn Valve Cartridge
Airflow Rate 1001 to 2500 mL/min.
Waterflow Rate 30 to 70 mL/min.