Learn More
Bochem™ Aluminum Lab Jacks
Product Code. 10313712
Click to view available options
Width (Metric):
100 mm
130 mm
200 mm
300 mm
Unit Size:
Each
Product Code. 10313712
Brand: Bochem™ 11022
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Material Aluminum, Stainless Steel
Dimensions (L x W) 160 x 130 mm
Length (Metric) Base 160 mm
Width (Metric) 130 mm
Includes Wheels
Product Type Lab Jack
Quantity 1 Ea.