Learn More
Bochem™ Aluminum Anodized Lab Jacks
Product Code. 10325911
Change view
Click to view available options
Width (Metric):
100 mm
130 mm
200 mm
240 mm
300 mm
400 mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) unitSize
10325911 100 mm Each
10653652 130 mm Each
10759611 200 mm Each
10294601 240 mm Each
10080911 300 mm Each
10616571 400 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10325911 Supplier Bochem™ Supplier No. 11016
Estimated Shipment: 20-10-2026
Add to basket
Add to basket
Manufactured according to DIN 12897 standards. Bochem™ aluminum Anodized Lab Jacks features a rounded, east to use ergonomic knob.
  • Improved scissor technology with reduced tolerances
  • Rounded, easy to use ergonomic knob
  • Scissor-construction made of 18/10-stainless steel
  • Maximal dynamic operating load (Kg max. dyn.) and the maximal static operating load (Kg max. stat.) norms are DIN 12897 compliant
  • Type 1 (240x240mm) uses a hand-operated adjusting wheel
  • Type 2 (300x300mm) includes ratchet, which connects to adjusting wheel
Specifications
Material Anodized Aluminum, Stainless Steel
Dimensions (L x W) 100 x 100 mm
Length (Metric) Base 100 mm
Width (Metric) 100 mm
Includes Wheels
Product Type Lab Jack
Quantity 1 Ea.