Learn More
Macherey-Nagel™ Alugram Aluminum Sheet SILGUR
Product Code. 12760833
Click to view available options
Dimensions (L x W):
10 x 20 cm
20 x 20 cm
Unit Size:
Pack of 20
Pack of 25
Product Code. 12760833
Brand: Macherey-Nagel™ 818423
Only null left
Specifications
Particle Size 5 to 17 μm
Pore Size 60 Å
Quantity 25 Pk.
Type Aluminum Sheet
Fluorescent Indicator UV254
Pore Volume 0.75 mL/g
Specific Surface ∼500 m2/g
Thickness (Metric) 0.20 mm
Dimensions (L x W) 20 x 20 cm