Learn More
Macherey-Nagel™ Alugram Aluminum Sheet CEL 300 UV254
Product Code. 10338651
Change view
Click to view available options
Dimensions (L x W):
5 x 2 cm
20 x 20 cm
Unit Size:
Pack of 25
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
10338651 20 x 20 cm Pack of 25
10516881 5 x 2 cm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10338651 Supplier Macherey-Nagel™ Supplier No. 818156
Only null left
Specifications
Quantity 25 Pk.
Type Aluminum Sheet
Fluorescent Indicator UV254
For Use With (Application) Partition Chromatography Of Polar Substances Such As Amino Acids, Carboxylic Acids Or Carbohydrates
Specific Surface 15,000 cm2/g
Dimensions (L x W) 20 x 20 cm