Learn More
Lenz Laborglasintrumente Alu Rings with Hooks
Product Code. p-7222461
Click to view available options
Size:
NS 19/26 to 24/29
NS 45/40
Unit Size:
Pack of 10
Product Code. 10643311
Brand: Lenz Laborglasintrumente 01102519
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • For cones and sockets to be used with springs
  • For the safe connection of conical ground joints
  • No breakage of glass hooks
Specifications
Quantity 10/Pk.
For Use With (Application) Cones and sockets to be used with springs
Size NS 19/26 to 24/29