Learn More
Tenak™ Alu FlexRack for Chest Freezers
Product Code. 11674742 Shop All TENAK Products
Click to view available options
Height (Metric):
340 mm
395 mm
505 mm
560 mm
615 mm
670 mm
Unit Size:
Each
Product Code. 11674742
Brand: Tenak™ TE20118
Only null left
Add to basket
Add to basket
Alu FlexRack is recommended for all applications where a flexible storage solution is needed.
  • Due to the adjustable shelves the rack can be used for all cryobox heights
  • Includes 1 shelf and 1 locking rod for securing the boxes
  • Constructed of aluminum
  • Can be used for any chest freezer
Specifications
Includes Locking rod
Material Aluminum
Shelves 1
For Use With (Equipment) Chest Frseezers
Width (Metric) 145 mm
Height (Metric) 560 mm
Depth (Metric) 140 mm