Learn More
Thermo Scientific™ ALPS 3000™ Automated Microplate Heat Sealer
GREENER_CHOICE
Product Code. 11557369
Change view
Click to view available options
Voltage:
110/240 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage unitSize
11557369 110/240 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11557369 Supplier Thermo Scientific™ Supplier No. AB3000
  • ONLINE EXCLUSIVE PRICE
    40420.00€
    37228.80€ / Pack of 1
    Save 3191.20€
    7% Off
View all promotions
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
Seal in heat fast with this automated heat sealer, designed for both optimal robotic integration in high throughput labs and manual benchtop control.
Quickly seal in heat with the Thermo Scientific™ ALPS 3000 Automated Microplate Heat Sealer. This compact sealer is designed for optimal robotic integration in high throughput labs, yet still allows for manual benchtop control.
Rapid Plate Sealing Action
  • Top-loaded seals are easily accessible
  • Seven available sealing options
  • Seals up to 6 plates per minute (10 secs/plate)
Fast, Easy to Use
  • Light (26 pounds) with a small footprint (165 × 405mm), takes up less precious bench space than comparable instruments
  • Available with a variety of available heat seals meet any laboratory application, including PCR, coulorimetric, fluorescence, long term storage, low temperature storage, piercing and resealing
  • On-board touch screen operation for benchtop use and and RS232 port with included ASCII programming parameters for robotic integration
Safe and Secure
  • Sensors detect the presence of sealing tape and plate
  • Air input allow for automatic system shutoff to eliminate product waste, system hold-ups and potential injury
TRUSTED_SUSTAINABILITY
Specifications
Hertz 50/60 Hz
Voltage 110/240 V
Description ALPS 3000 Automated Microplate Heat Sealer
Dimensions (L x W x H) 16.3 x 7.3 x 13.8 in. (414 x 185 x 350 mm)
Electrical Requirements 110/240 V, 50/60 Hz
Height (English) 13.8 in.
Height (Metric) 350 mm
Length (English) 16.3 in.
Length (Metric) 414 mm
No. per Pack 1 Ea.
Sealing Temperature Range 200°C
Weight (English) 26.5 lb.
Weight (Metric) 12 kg
Width (English) 7.3 in.
Width (Metric) 185 mm
Product Type Plate Sealer