missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 18 in. Solvex™ 37-185 Nitrile Gloves

Product Code. 12669701 Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
11
Unit Size:
1 pair
12 pairs
This item is not returnable. View return policy

Product Code. 12669701

Brand: Ansell 37185_7

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Heavy-duty, elbow length nitrile chemical resistant gloves suitable for a wide range of applications.

  • High performance nitrile compound
  • Combination of chemical resistance and strength ,for optimal results in wet or dry environments
  • Snag, puncture and abrasion protection compared with rubber or neoprene gloves
  • High levels of flexibility, comfort and dexterity
  • Suitable for heavy-duty cleaning applications
  • Complies with FDA food handling requirements (21 CFR 177.2600)
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance FDA 21 CFR 177.2600
Certifications/Compliance (Europe) CE 0493, EN 420:2003 + A1:2009, Category III
Chlorinated Yes
Applications Industrial
Color Green
Latex Free Yes
Material Nitrile
Product Type Chemical-resistant Gloves
Size 7
Thickness (English) 22 mil
Thickness (Metric) 0.55 mm
EN388 Standard EN 388:4102X, EN 388:4102
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AGJKLPT
EN ISO 374 - 5 Bacteria, Fungi only
For Use With (Application) Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
Glove Cuff Style Gauntlet
Grip Design Sandpatch
Lining Unlined
Manufacturing/QMS Audit Standards ISO 18889
Model 37-185 7 117298
Series AlphaTec™ Solvex™ 37-185
Length (English) 18 in.
Length (Metric) 45.7 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.