Learn More
Ansell™ AlphaTec 53-001 Gloves
Shop All Ansell Products
Click to view available options
:
Size: 6; Color: White; Material: Nylon
Description
- These acid-resistant gloves offer a nitrile and neoprene barrier that provides advanced chemical protection from acids to hydrocarbons and beyond
- These AlphaTec™ 53-001 black chemical resistant gloves have ‘Type A’ chemical protection under EN ISO 374:2016
- MICROCHEM™ Chemical Barrier Technology combines polymers in proprietary blends or multi-layer structures specific to their application, providing superior protection to wearers
- ANSELL GRIP™ Technology for the handling of wet or oily parts providing enhanced dexterity, grip and comfort
Specifications
Specifications
| Color | White, Black |
| Latex Free | Yes |
| Material | Nylon |
| Size | 6 |
| Style | 53-001 |
| Length (Metric) | 350 mm |