Learn More
Ansell™ AlphaTec™ 4000 Ultrasonically Welded and Taped
Product Code. p-200004969
Click to view available options
Size:
Small
Medium
Large
2X-Large
3X-Large
Unit Size:
Pack of 10
Product Code. 17130645
Brand: Ansell™ GR40T0012102
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
AlphaTec™ 4000 Ultrasonically welded and taped consists of a unique multi-layer barrier fabric renowned for its lightweight, yet robust textile feel and exceptional barrier to organic and inorganic chemicals.
Consists of a unique multi-layer barrier fabric that provides lightweight and robust textile feel with exceptional barrier to organic and inorganic chemicals.
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Designed to protect, typical innovative features include dual zip systems and double cuffs
  • Welded and taped seams, our highest barrier to liquids and particulates
  • Enhanced comfort, knitted inner cuffs and inner textile type fabric improves wearer acceptance
  • Anti-static, tested according to EN 1149-5
TRUSTED_SUSTAINABILITY
Specifications
Product Type Chemical Protective Clothing
Size Small
Gender Unisex
Material Multi-layer Barrier Laminate Fabric
Color Spruce Green
Disposable Reusable
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic, Double Inner Knit
Neck Style Hooded
Hood Type 2-panel, Elastic
Closure Type Double Zipper
Seam Type Welded, Taped
Sterility Non-sterile
Protection Class CE CATEGORY III
Resistance Chemicals, Particle
Construction 2-piece Hood, Elasticated Waist, Double Cuffs and Ankles, Double Zip Closure, Double Cuffs with Knitted Inner Cuff, Attached Ansell Barrier Gloves
Collar Type Hood
Certifications/Compliance (Europe) CE CATEGORY III (Type 3-B, Type 4-B, Type 5-B, EN 1073-2, EN 14126, EN 1149-5)
No. of Pockets None
For Use With (Application) Contaminated Environments, Chemical Handling, Filling, blending and charging of raw materials, Transferring liquids and solids, Opening and draining pumps valves or lines, Loading and unloading of proces equipment, Raw material sample collection, Concentration mixing and palletisation, Hazmat response, Assembly and disassembly of large pipes