Learn More
Ansell™ AlphaTec™ 4000 Ultrasonically Welded and Taped
Click to view available options
Size:
Small
Medium
Large
2X-Large
3X-Large
Unit Size:
Pack of 10
Description
Consists of a unique multi-layer barrier fabric that provides lightweight and robust textile feel with exceptional barrier to organic and inorganic chemicals.
- Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
- Designed to protect, typical innovative features include dual zip systems and double cuffs
- Welded and taped seams, our highest barrier to liquids and particulates
- Enhanced comfort, knitted inner cuffs and inner textile type fabric improves wearer acceptance
- Anti-static, tested according to EN 1149-5
Specifications
Specifications
| Product Type | Chemical Protective Clothing |
| Size | Medium |
| Gender | Unisex |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | Spruce Green |
| Disposable | Reusable |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Elastic, Double Inner Knit |
| Neck Style | Hooded |
| Show More |