missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec 4000 AVANT AIRline
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Includes:
Attached Socks and Gloves
Socks and Elasticated Boot Overlap
Socks and Neoprene Gloves
Style:
G40T755
G40T755B
G40T755C
G40T755D
Unit Size:
Pack of 3
Description
- Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
- Panoramic visor design ensures a good field of vision for the wearer
- A range of sleeve options available, including double cuffs and permanently attached chemical protective gloves.
- HEPA filter provides secondary protection from airline contamination and a SMC silencer ensures the noise inside the suit is always below 70 dB (even at maximum airflow).
- Suit/Belt mounted regulator complete with a low flow warning whistle the AVANT STS regulator can be mounted on the suit and removed for reuse providing cleaning and decontamination is permissible.
- Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer
- Attached socks with elasticated boot overflap - Socks are designed to be worn inside chemical protective boots with the leg overflap worn outside to reduce the potential for chemical ingress.
Specifications
Specifications
| Color | Green |
| For Use With (Application) | Contaminated Environments, Chemical Handling, Filling, blending and charging of raw materials, Transferring liquids and solids, Opening and draining pumps valves or lines, Loading and unloading of proces equipment, Raw material sample collection, Concentration mixing and palletisation, Hazmat response, Assembly and disassembly of large pipes |
| Hood Type | Fully Encapsulated |
| Material | Multi-layer Barrier Laminate Fabric |
| Size | 2X-Large |
| Includes | Socks and Elasticated Boot Overlap |
| Product Type | Encapsulated chemical protective suits for use with air-fed/supplied systems |
| Seam Type | Welded and Taped |
| Style | G40T755 |
| Weight (Metric) | 103 gsm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction