Learn More
Ansell AlphaTec™ 2000 STANDARD Oversleeves Bound
Product Code. p-7228379
Click to view available options
Size:
Universal
X-Large
Unit Size:
250 pairs
300 pairs
Product Code. 16854800
Brand: Ansell WH20B0060800
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Comfortable oversleeves featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress.
  • Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
  • Optimized fit, improving wearer comfort and safety
  • Low Linting
TRUSTED_SUSTAINABILITY
Specifications
Size Universal
Material Microporous Non-Woven Polyethylene Laminate
Color White
Seam Type Bound
Model WH20B
For Use With (Application) Cleanroom Cleaning and Preparing, Production Line Support & Maintenance, Maintenance of Plant and Machines, Veterinary Services
Certifications/Compliance (Europe) EN 13034:2005 + A1:2009, EN 14126:2003, EN ISO 13688:2013, Partial Body Protection Only
Weight (Metric) 63 gsm