Learn More
Ansell AlphaTec™ 2000 STANDARD Lab Coat Bound
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 60
Description
Designed to allow perspiration to escape from the suit and withstand saturation of liquid chemicals.
- Filter 100% of particulates down to 0.01 microns in size
- Proven protective barrier to low concentration liquid chemicals, liquids, and particulate biological hazards
- Comfortable to wear featuring moisture vapor permeable fabric to help reduce the risk of heat stress
- Optimized fit, improving wearer comfort and safety
- Low linting
Specifications
Specifications
| Product Type | Controlled Environments Lab Coat |
| Size | Medium |
| Gender | Unisex |
| Material | Microporous Non-woven Polyethylene Laminate |
| Color | White |
| Closure Type | Zipper |
| Cuff Style | Elastic |
| Disposable | Reusable |
| Resistance | Fluids |
| Style | Knee-length |
| Show More |