Learn More
Ansell AlphaTec™ 2000 STANDARD Apron with Sleeves Bound - Model 214
Product Code. p-7239439 Shop All Ansell Products
Click to view available options
Size:
Medium
Large
Unit Size:
Pack of 60
Product Code. 16190551
Brand: Ansell WH20B0021403
Only null left
Add to basket
Add to basket
AlphaTec™ Apron with Sleeves Bound - Superior breathable microporous laminate technology
  • Proven protective barrier to low concentration liquid chemicals, liquid & particulate biological hazards
  • Silicone Free
  • Low Linting
TRUSTED_SUSTAINABILITY
Specifications
Type Apron
Size Medium
Material Microporous Laminate
Color White
Disposable Limited-use
Closure Type Hook and Loop (Rear)
Sterility Non-sterile
For Use With (Application) Critical and Controlled Environments
Style AlphaTec 2000-214
Seam Type Bound
Certifications/Compliance (Europe) EN 14126