Learn More
Ansell AlphaTec™ 1800 Ts PLUS
Product Code. 16834740
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Style:
W18T111
W18T122
W18T156
Unit Size:
Pack of 30
Pack of 35
Pack of 40
Pack of 45
Product Code. 16834740
Brand: Ansell WH18T0012205
Lightweight coveralls with breathable microporous laminate technology and Type 4/5/6 protection.
  • 3 piece hood
  • Elasticated hood, wrists, waist and ankles
  • 2-way front zipper with resealable storm flap
  • Finger Loops
  • Chinstrap
TRUSTED_SUSTAINABILITY
Specifications
Size X-Large
Material Microporous Polyethylene Laminate (Non-woven)
Color White
Seam Type Stitched, Taped
Certifications/Compliance (Europe) EN 374:2003, EN 14605:2005 + A1:2009,EN ISO 13982-1:2004 + A1:2010,EN 13034:2005 + A1:2009,EN 1073-2:2002,EN 1149-5:2008,EN 14126:2003
Includes Hood and Boots
For Use With (Application) Clean Room Cleaning and Preparing, Body Painting Including Surface Inspection, Production Line Support & Maintenance, Cleaning of Plant and Machines, Maintenance of Plant and Machines, Fibre Glass Resin Applications, Handling of Powders
Weight (Metric) 53 gsm
Style W18T122