missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 1800 Ts PLUS
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Style:
W18T111
W18T122
W18T156
Unit Size:
Pack of 30
Pack of 35
Pack of 40
Pack of 45
Description
- 3 piece hood
- Elasticated hood, wrists, waist and ankles
- 2-way front zipper with resealable storm flap
- Finger Loops
- Chinstrap
Specifications
Specifications
| Size | Medium |
| Material | Microporous Polyethylene Laminate (Non-woven) |
| Color | White |
| Seam Type | Stitched, Taped |
| Includes | Hood and Boots |
| For Use With (Application) | Clean Room Cleaning and Preparing, Body Painting Including Surface Inspection, Production Line Support & Maintenance, Cleaning of Plant and Machines, Maintenance of Plant and Machines, Fibre Glass Resin Applications, Handling of Powders |
| Weight (Metric) | 53 gsm |
| Style | W18T122 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction