Description
- Lightweight microporous polyethylene (PE) laminate
- Bound seam for additional protection and strength
- 2-way front zipper with re-sealable storm flap
- Thumb loop ensures arm protection remains in place when arm is raised
- Elasticated waist to enhance fit
- Innovative three-piece elasticated hood construction provides added comfort, durability and fit
- Elasticated ankles and wrists
Specifications
Specifications
| Product Type | Chemical-resistant Coveralls |
| Size | X-Large |
| Gender | Unisex |
| Material | Polyethylene, Polypropylene |
| Color | White |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Show More |