Learn More
Skytec™ Alphashield 2000+- S2BH+ Disposable Coverall
Description
- Spun bonded polypropylene with outer breathable film with chemical liquid (EN368 and blood (ASTM F1670) penetration resistance for premium protection
- Strong, durable, and comfortable
- Improved fit with two-piece crotch pattern
- Quick and easy donning/doffing in an emergency with longer 2-way zip
- Fully elasticated hood, ankles, and wrists
Specifications
Specifications
| Product Type | Chemical-resistant Coveralls |
| Size | Medium |
| Gender | Unisex |
| Material | Polypropylene |
| Color | White |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Show More |