Learn More
Alpha5IA, TRC
Specifications
Specifications
| Chemical Name or Material | Alpha5IA |
| CAS | 215874-86-5 |
| Molecular Formula | C17H14N8O2 |
| Quantity | 10 mg |
| Synonym | 3-(5-Methyl-3-isoxazolyl)-6-[(1-methyl-1H-1,2,3-triazol-4-yl)methoxy]-1,2,4-triazolo[3,4-a]phthalazine |
| InChI Formula | InChI=1S/C17H14N8O2/c1-10-7-14(22-27-10)16-20-19-15-12-5-3-4-6-13(12)17(21-25(15)16)26-9-11-8-24(2)23-18-11/h3-8H,9H2,1-2H3 |
| SMILES | Cc1onc(c1)c2nnc3c4ccccc4c(OCc5cn(C)nn5)nn23 |
| IUPAC Name | 5-methyl-3-[6-[(1-methyltriazol-4-yl)methoxy]-[1,2,4]triazolo[3,4-a]phthalazin-3-yl]-1,2-oxazole |
| Molecular Weight (g/mol) | 362.35 |
| Formula Weight | 362.124 |