Learn More
STEMCELL Technologies Alpha MEM with Nucleosides
Description
- Minimum Essential Medium Eagle - Alpha Modification (Alpha MEM) with Nucleosides is recommended for a wide variety of cell culture applications
- Selection of a suitable nutrient medium is dependent on type of cell, conditions of culture, and degree of chemical definition
- MEM has typically been used for the cultivation of cells grown in monolayers, however additional supplementation to Alpha MEM has expanded the applications for this medium
- MEM was developed by Harry Eagle when Basal Medium Eagle (BME) was found not adequate to support the growth of HeLa cells
- It was discovered that BME could support the growth of a wider variety of cells with the addition of certain medium supplements (e.g. higher concentrations of amino acids), which were incorporated into the MEM formulation
- Alpha MEM includes the non-essential amino acids, sodium pyruvate, lipoic acid, ascorbic acid, biotin, and vitamin B12
Specifications
Specifications
| For Use With (Application) | Cell Culture |
| Quantity | 500 mL |