missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals alpha-Mating Factor (1-6)

Product Code. p-7076676
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
This item is not returnable. View return policy

Product Code. 15457859

Brand: Thermo Scientific Chemicals J66484.MA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 65418-88-4 | C45H59N11O8 | 882.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65418-88-4

Specifications

Content And Storage Store at -20°C
Format Solid
Molecular Formula C45H59N11O8
InChI Key PBLMMVDDGREFKH-UHFFFAOYNA-N
SMILES CC(C)CC(NC(=O)C(CCC(N)=O)NC(=O)C(CC(C)C)NC(=O)C(CC1=CNC2=CC=CC=C12)NC(=O)C(CC1=CN=CN1)NC(=O)C(N)CC1=CNC2=CC=CC=C12)C(O)=O
IUPAC Name 2-{2-[2-(2-{2-[2-amino-3-(1H-indol-3-yl)propanamido]-3-(1H-imidazol-5-yl)propanamido}-3-(1H-indol-3-yl)propanamido)-4-methylpentanamido]-4-carbamoylbutanamido}-4-methylpentanoic acid
Quantity 10 mg
Molecular Weight (g/mol) 882.04

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.