Learn More
Alpha-Hydroxy Trimethoprim (Impurity), TRC
Specifications
Specifications
| Chemical Name or Material | (RS)-(2,4-Diaminopyrimidin-5-yl)(3,4,5-trimethoxyphenyl)methanol |
| CAS | 29606-06-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C14 H18 N4 O4 |
| Quantity | 25 mg |
| Synonym | (RS)-(2,4-Diaminopyrimidin-5-yl)(3,4,5-trimethoxyphenyl)methanol, Trimethoprim Imp. C (EP) |
| InChI Formula | InChI=1S/C14H18N4O4/c1-20-9-4-7(5-10(21-2)12(9)22-3)11(19)8-6-17-14(16)18-13(8)15/h4-6,11,19H,1-3H3,(H4,15,16,17,18) |
| SMILES | COc1cc(cc(OC)c1OC)C(O)c2cnc(N)nc2N |
| IUPAC Name | (2,4-diaminopyrimidin-5-yl)-(3,4,5-trimethoxyphenyl)methanol |
| Molecular Weight (g/mol) | 306.32 |
| Show More |