missing translation for 'onlineSavingsMsg'
Learn More

alpha-D-Glucopyranoside-beta-D-fructofuranosyl Octadecanoate, TRC

Product Code. 30394692
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394692 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394692 Supplier Toronto Research Chemicals Supplier No. TRCG59664525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material alpha-D-Glucopyranoside-beta-D-fructofuranosyl Octadecanoate
CAS 37318-31-3
Molecular Formula C30 H56 O12
Quantity 25 mg
Synonym α-D-Glucopyranoside β-D-fructofuranosyl octadecanoate, alpha-D-Glucopyranoside-beta-D-fructofuranosyl Octadecanoate, Sucrose, stearate (6CI), Cane sugar stearate, Cosmelike S 10, Cosmelike S 110, Crodesta F 110, Crodesta F 140, Crodesta F 160, Crodesta F 90, DK 160, DK 50, DK Ester O 10, DKF 10, DUB-SE 11S, Nitto Ester S 370, Rikemal A, Ryoto S 1170, Ryoto S 170, Ryoto Sugar Ester S 070, Ryoto Sugar Ester S 1170, Ryoto Sugar Ester S 1170F, Ryoto Sugar Ester S 1170S, Ryoto Sugar Ester S 1570, Ryoto Sugar Ester S 170, Ryoto Sugar Ester S 270, Ryoto Sugar Ester S 370F, Ryoto Sugar Ester S 395, Ryoto Sugar Ester S 570, Ryoto Sugar Ester S 70, Ryoto Sugar Ester S 970, Ryoto Sugar Ester SM 1800, S 070, S 1170, S 1570, S 170, S 270, S 395, S 970, SE 1570, SE 970, SEFA polystearate, SFE 1815, SM 90, SM 90 (sugar ester), SP 10C, SS 15, SS 7, Saccharose stearate, Sefose 1618H, Sisterna SP 50, Sucroester WE 11, Sucrose octadecanoate, Sucrose stearate, Surfhope C 1816, Surfhope J 1809, Surfhope J 1811, Surfhope J 1815, Surfhope SE COSME C 1800, Surfhope SE COSME C 1801, Surfhope SE COSME C 1802, Surfhope SE Cosme C 1811, Surfhope SE Cosme C 1815, Surfhope SE Cosme C 1816, Surfhope Sepharma J 1811F
InChI Formula InChI=1S/C30H56O12/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-23(33)42-30(20-32)28(38)25(35)22(41-30)19-39-29-27(37)26(36)24(34)21(18-31)40-29/h21-22,24-29,31-32,34-38H,2-20H2,1H3/t21-,22-,24-,25-,26+,27-,28+,29+,30+/m1/s1
SMILES CCCCCCCCCCCCCCCCCC(=O)O[C@]1(CO)O[C@H](CO[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
IUPAC Name [(2S,3S,4S,5R)-3,4-dihydroxy-2-(hydroxymethyl)-5-[[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxolan-2-yl] octadecanoate
Molecular Weight (g/mol) 608.758
Formula Weight 608.377
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.