missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-D-Glucopyranoside-beta-D-fructofuranosyl Octadecanoate, TRC
Specifications
Specifications
| Chemical Name or Material | alpha-D-Glucopyranoside-beta-D-fructofuranosyl Octadecanoate |
| CAS | 37318-31-3 |
| Molecular Formula | C30 H56 O12 |
| Quantity | 50 mg |
| Synonym | α-D-Glucopyranoside β-D-fructofuranosyl octadecanoate, alpha-D-Glucopyranoside-beta-D-fructofuranosyl Octadecanoate, Sucrose, stearate (6CI), Cane sugar stearate, Cosmelike S 10, Cosmelike S 110, Crodesta F 110, Crodesta F 140, Crodesta F 160, Crodesta F 90, DK 160, DK 50, DK Ester O 10, DKF 10, DUB-SE 11S, Nitto Ester S 370, Rikemal A, Ryoto S 1170, Ryoto S 170, Ryoto Sugar Ester S 070, Ryoto Sugar Ester S 1170, Ryoto Sugar Ester S 1170F, Ryoto Sugar Ester S 1170S, Ryoto Sugar Ester S 1570, Ryoto Sugar Ester S 170, Ryoto Sugar Ester S 270, Ryoto Sugar Ester S 370F, Ryoto Sugar Ester S 395, Ryoto Sugar Ester S 570, Ryoto Sugar Ester S 70, Ryoto Sugar Ester S 970, Ryoto Sugar Ester SM 1800, S 070, S 1170, S 1570, S 170, S 270, S 395, S 970, SE 1570, SE 970, SEFA polystearate, SFE 1815, SM 90, SM 90 (sugar ester), SP 10C, SS 15, SS 7, Saccharose stearate, Sefose 1618H, Sisterna SP 50, Sucroester WE 11, Sucrose octadecanoate, Sucrose stearate, Surfhope C 1816, Surfhope J 1809, Surfhope J 1811, Surfhope J 1815, Surfhope SE COSME C 1800, Surfhope SE COSME C 1801, Surfhope SE COSME C 1802, Surfhope SE Cosme C 1811, Surfhope SE Cosme C 1815, Surfhope SE Cosme C 1816, Surfhope Sepharma J 1811F |
| InChI Formula | InChI=1S/C30H56O12/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-23(33)42-30(20-32)28(38)25(35)22(41-30)19-39-29-27(37)26(36)24(34)21(18-31)40-29/h21-22,24-29,31-32,34-38H,2-20H2,1H3/t21-,22-,24-,25-,26+,27-,28+,29+,30+/m1/s1 |
| SMILES | CCCCCCCCCCCCCCCCCC(=O)O[C@]1(CO)O[C@H](CO[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O |
| IUPAC Name | [(2S,3S,4S,5R)-3,4-dihydroxy-2-(hydroxymethyl)-5-[[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxolan-2-yl] octadecanoate |
| Molecular Weight (g/mol) | 608.758 |
| Formula Weight | 608.377 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?