Learn More
Alpha-Cyclogeranonitrile, TRC
Specifications
Specifications
| Chemical Name or Material | alpha-Cyclogeranonitrile |
| CAS | 57524-13-7 |
| Molecular Formula | C10H15N |
| Quantity | 250 mg |
| Synonym | 2,6,6-Trimethyl-2-cyclohexene-1-carbonitrile |
| InChI Formula | InChI=1S/C10H15N/c1-8-5-4-6-10(2,3)9(8)7-11/h5,9H,4,6H2,1-3H3 |
| SMILES | CC1=CCCC(C)(C)C1C#N |
| IUPAC Name | 2,6,6-trimethylcyclohex-2-ene-1-carbonitrile |
| Molecular Weight (g/mol) | 149.23 |
| Formula Weight | 149.1204 |