Learn More
Alpha,Alpha'-Dilaurin, TRC
Product Code. 30376447
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376447 1 g -
30370635 250 mg 250mg
30370411 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30376447 Supplier Toronto Research Chemicals Supplier No. TRCD4599201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 539-93-5
Molecular Formula C27H52O5
Molecular Weight (g/mol) 456.7
Synonym Dodecanoic Acid 2-Hydroxy-1,3-propanediyl Ester, 1,3-Dilaurin, 1,3-Dilauroylglycerol, Glycerol 1,3-didodecanoate, Glycerol 1,3-dilaurate
IUPAC Name (3-dodecanoyloxy-2-hydroxypropyl) dodecanoate
SMILES CCCCCCCCCCCC(=O)OCC(O)COC(=O)CCCCCCCCCCC
Specifications
Quantity 1 g
InChI Formula InChI=1S/C27H52O5/c1-3-5-7-9-11-13-15-17-19-21-26(29)31-23-25(28)24-32-27(30)22-20-18-16-14-12-10-8-6-4-2/h25,28H,3-24H2,1-2H3
Formula Weight 456.3815
Chemical Name or Material alpha,alpha'-Dilaurin