Learn More
Alpha,Alpha,Alpha’,Alpha’-(Tetramethyl-d12)-5-bromomethyl-1,3-benzenediacetonitrile, TRC
Specifications
Specifications
| Chemical Name or Material | alpha,alpha,alpha',alpha'-(Tetramethyl-d12)-5-bromomethyl-1,3-benzenediacetonitrile |
| CAS | 1185016-59-4 |
| Molecular Formula | C15H5D12BrN2 |
| Quantity | 10 mg |
| Synonym | 5-(Bromomethyl)-alpha1,alpha1,alpha3,alpha3-tetra(methyl-d3)-1,3-benzenediacetonitrile, 3,5-Bis(1-cyano-1-methylethyl)benzyl-d12 Bromide, 3,5-Bis(2-cyanoprop-2-yl)benzyl-d12 Bromide, 2,2’-(5-Bromomethyl)-1,3-phenylene)-bis(methylpropionitrile)-d12 |
| InChI Formula | InChI=1S/C15H17BrN2/c1-14(2,9-17)12-5-11(8-16)6-13(7-12)15(3,4)10-18/h5-7H,8H2,1-4H3/i1D3,2D3,3D3,4D3 |
| SMILES | [2H]C([2H])([2H])C(C#N)(c1cc(CBr)cc(c1)C(C#N)(C([2H])([2H])[2H])C([2H])([2H])[2H])C([2H])([2H])[2H] |
| IUPAC Name | 2-[3-(bromomethyl)-5-(2-cyano-1,1,1,3,3,3-hexadeuteriopropan-2-yl)phenyl]-3,3,3-trideuterio-2-(trideuteriomethyl)propanenitrile |
| Molecular Weight (g/mol) | 317.29 |
| Formula Weight | 316.1328 |