Learn More
alpha 2u-Globulin Goat anti-Rat, Polyclonal, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  R&D Systems AF586-SP
Encompass
Product Code. 15708872
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
alpha 2u-Globulin Polyclonal specifically detects alpha 2u-Globulin in Rat samples. It is validated for Western Blot, Immunohistochemistry.