Learn More
IKA™ Alnico V Magnetic Stirring Bars
Product Code. 15398143 Shop All IKA Products
Click to view available options
Length (Metric):
10 mm
15 mm
25 mm
30 mm
50 mm
For Use With (Equipment):
Flat bottom containers
Mixing Reagents
Unit Size:
Each
Pack of 5
Product Code. 15398143
Brand: IKA™ 0004488700
Specifications
For Use With (Equipment) Flat bottom containers
Coating PTFE
Diameter (Metric) 6 mm
Length (Metric) 15 mm
Material Alnico Magnet
Quantity 5/Pk.
Shape Sphere, Cylinder
Type Magnetic Stir Bar