Learn More
Alloy Steel, Welding Wire, D6-AC, K24728, 100g, ARMI∣MBH
Description
Alloy Steel, Welding Wire, D6-AC, K24728, 100g
Specifications
Specifications
| Chemical Name or Material | Boron/Nitrogen/Arsenic/Titanium/Cobalt/Tin/Niobium/Phosphorus/Tungsten/Aluminium/Copper/Vanadium/Silicon/Carbon/Nickel/Manganese/Molybdenum/Chromium/Calcium/Lead/Zirconium/Antimony/Oxygen |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Boron 0.0003 %/Nitrogen 0.0028 %/Arsenic 0.003 %/Titanium 0.0038 %/Cobalt 0.0054 %/Tin 0.0055 %/Niobium 0.006 %/Phosphorus 0.008 %/Tungsten 0.01 %/Aluminium 0.092 %/Copper 0.1 %/Vanadium 0.12 %/Silicon 0.22 %/Carbon 0.469 %/Nickel 0.57 %/Manganese 0.7 %/Molybdenum 0.99 %/Chromium 1.03 %/Calcium (0.001) %/Lead (0.001) %/Zirconium (0.001) %/Antimony (0.002) %/Oxygen (0.002) % |