missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, Reinforcing Steel, A706 Grade 60, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Alloy Steel, Reinforcing Steel, A706 Grade 60, Ø32x19mm
Specifications
Specifications
| Chemical Name or Material | Nitrogen/Niobium/Tin/Cobalt/Phosphorus/Aluminium/Nickel/Copper/Manganese/Carbon/Vanadium/Silicon/Molybdenum/Chromium/Cadmium/Magnesium/Calcium/Lead/Antimony/Bismuth/Zirconium/Tantalum/Selenium/Zinc/Boron/Oxygen/Arsenic/Titanium/Tungsten |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Nitrogen 0.005 %/Niobium 0.006 %/Tin 0.006 %/Cobalt 0.009 %/Phosphorus 0.01 %/Aluminium 0.032 %/Nickel 0.062 %/Copper 0.073 %/Manganese 0.317 %/Carbon 0.389 %/Vanadium 0.515 %/Silicon 0.91 %/Molybdenum 1.3 %/Chromium 4.82 %/Cadmium (<0.0001) %/Magnesium (<0.0001) %/Calcium (<0.0003) %/Lead (<0.0005) %/Antimony (<0.002) %/Bismuth (<0.004) %/Zirconium (<0.005) %/Tantalum (<0.008) %/Selenium (<0.011) %/Zinc (<0.011) %/Boron (0.0004) %/Oxygen (0.002) %/Arsenic (0.004) %/Titanium (0.0048) %/Tungsten (0.007) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction