missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, Ni/Cr/Mo, AISI 86L20, G86200, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Alloy Steel, Ni/Cr/Mo, AISI 86L20, G86200, 100g
Specifications
Specifications
| Chemical Name or Material | Vanadium/Nitrogen/Cobalt/Phosphorus/Tin/Sulfur/Aluminium/Carbon/Lead/Molybdenum/Copper/Silicon/Nickel/Chromium/Manganese |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Vanadium 0.0016 %/Nitrogen 0.0052 %/Cobalt 0.0085 %/Phosphorus 0.009 %/Tin 0.014 %/Sulfur 0.015 %/Aluminium 0.025 %/Carbon 0.175 %/Lead 0.2 %/Molybdenum 0.203 %/Copper 0.242 %/Silicon 0.29 %/Nickel 0.458 %/Chromium 0.53 %/Manganese 0.794 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction