missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Alloy Steel, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Phosphorus/Tin/Titanium/Aluminium/Copper/Tungsten/Niobium/Cobalt/Molybdenum/Vanadium/Carbon/Nickel/Chromium/Silicon/Manganese |
| CAS | 7439-89-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Phosphorus (0.068) %/Tin (0.07) %/Titanium (0.09) %/Aluminium (0.092) %/Copper (0.11) %/Tungsten (0.125) %/Niobium (0.13) %/Cobalt (0.31) %/Molybdenum (0.37) %/Vanadium (0.37) %/Carbon (0.39) %/Nickel (0.5) %/Chromium (0.64) %/Silicon (1) %/Manganese (1.25) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction