Learn More
Tocris Bioscience™ Allopregnanolone
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Allopregnanolone is a GABAA receptor positive allosteric modulator. Neuroactive steroid.
Specifications
Specifications
| Chemical Name or Material | (3α,5α)-3-Hydroxy-pregnan-20-one |
| CAS | 516-54-1 |
| Quantity | 10 mg |
| Target | GABA-A Receptor Modulators |
| Molecular Formula | C21H34O2 |