Learn More
Allo-3Alpha-tetrahydro Cortisol, TRC
Product Code. 30136020
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136020 50 mg 50mg
30004920 10 mg 10mg
30010967 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136020 Supplier Toronto Research Chemicals Supplier No. TRCA54850050MG
 Restricted Item
Estimated Shipment: 10-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 302-91-0
Molecular Formula C21H34O5
Molecular Weight (g/mol) 366.49
Synonym Pregnan-20-one, 3,11,17,21-tetrahydroxy-, (3α,5α,11β)-, 5α-Pregnan-20-one, 3α,11β,17,21-tetrahydroxy- (8CI), (3α,5α,11β)-3,11,17,21-Tetrahydroxypregnan-20-one, 3α,11β,17,21-Tetrahydroxy-5α-pregnan-20-one, 3α,11β,17α,21-Tetrahydroxy-5α-pregnan-20-one, 3α,5α-Tetrahydrocortisol, 5α-Pregnane-3α,11β,17α,21-tetraol-20-one, 5α-Pregnane-3α,11β,17α,21-tetrol-20-one, 5α-THF, 5α-Tetrahydrocortisol, AL 3308, ATHF, Allo-3α-tetrahydrocortisol, Allopregnane-3α,11β,17α,21-tetrol-20-one, Allotetrahydrocompound F, Allotetrahydrocortisol, Cortisol, allotetrahydro-, Kendall's compound C, NSC 86469, Reichstein's substance C, Wintersteiner's compound D, allo-Tetrahydrocortisol, α-THF
SMILES C[C@]12CC[C@@H](O)C[C@@H]1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)CO)[C@@]4(C)C[C@H](O)[C@H]23
Specifications
Quantity 50 mg
Formula Weight 366.2406
Percent Purity >95%
Chemical Name or Material Allo-3alpha-tetrahydro Cortisol
Grade HPLC