Learn More
Allihn Condenser with Ground Cone and Socket
Product Code. 15507790
Click to view available options
Standard Taper Joint No:
24/29
29/32
Length (Metric) Jacket:
545 mm
550 mm
Unit Size:
Each
Pack of 2
Product Code. 15507790
Brand: Quickfit CX7/44/SC
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Ideal for reflux applications.
  • Inner bulb sections provide increased cooling surface
  • With detachable plastic screwthread connectors for easy and safe attachment of flexible tubing
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) Jacket 550 mm
Quantity 1 Each
Standard Taper Joint No 29/32
Material Borosilicate Glass
Type Allihn Condenser